Product Details

SNP ID
rs782791
Assay Type
Validated
NCBI dbSNP Submissions
72
Location
Chr.1:203029691 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGAGATATTTAACCTGGAGCAGC[A/G]AAGCCTCTGGGGACCCAGGACACTG
Phenotype
MIM: 603145
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPFIA4 PubMed Links

Gene Details

Gene
PPFIA4
Gene Name
PTPRF interacting protein alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304331.1 Intron NP_001291260.1
NM_001304332.1 Intron NP_001291261.1
XM_006711586.2 Intron XP_006711649.1
XM_011510070.2 Intron XP_011508372.1
XM_017002606.1 Intron XP_016858095.1
XM_017002607.1 Intron XP_016858096.1
XM_017002608.1 Intron XP_016858097.1
XM_017002609.1 Intron XP_016858098.1
XM_017002610.1 Intron XP_016858099.1
Gene
TMEM183A
Gene Name
transmembrane protein 183A
There are no transcripts associated with this gene.

View Full Product Details