Product Details

SNP ID
rs1498195
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76061834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCCTACCCTCCTTGCTGCCCT[G/T]CACACTATCTTCTGGTCTCACTGCC
Phenotype
MIM: 610894
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
NRG4 PubMed Links

Gene Details

Gene
NRG4
Gene Name
neuregulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138573.3 1164 Intron NP_612640.1
XM_017021937.1 1164 Intron XP_016877426.1
XM_017021938.1 1164 Intron XP_016877427.1
XM_017021939.1 1164 Intron XP_016877428.1
XM_017021940.1 1164 Intron XP_016877429.1
XM_017021941.1 1164 Intron XP_016877430.1
XM_017021942.1 1164 Intron XP_016877431.1
XM_017021943.1 1164 Intron XP_016877432.1
XM_017021944.1 1164 Intron XP_016877433.1
XM_017021945.1 1164 Intron XP_016877434.1
XM_017021946.1 1164 Intron XP_016877435.1
XM_017021947.1 1164 Intron XP_016877436.1
XM_017021948.1 1164 UTR 5 XP_016877437.1
Gene
TMEM266
Gene Name
transmembrane protein 266
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152335.2 1164 Intron NP_689548.2
XM_005254160.2 1164 Intron XP_005254217.1
XM_017021915.1 1164 Intron XP_016877404.1
XM_017021916.1 1164 Intron XP_016877405.1
XM_017021917.1 1164 Intron XP_016877406.1

View Full Product Details