Product Details

SNP ID
rs1044797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:63121975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGGCCATGAAAAGTTTAGATAT[A/G]AAGAAGGCAGTTTTCTTTTTTTCTT
Phenotype
MIM: 613580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DBIL5P2 PubMed Links

Gene Details

Gene
DBIL5P2
Gene Name
diazepam binding inhibitor-like 5 pseudogene 2
There are no transcripts associated with this gene.

Gene
WDPCP
Gene Name
WD repeat containing planar cell polarity effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042692.2 2558 UTR 3 NP_001036157.1
NM_015910.5 2558 UTR 3 NP_056994.3
XM_005264348.3 2558 Missense Mutation TCA,TTA S731L XP_005264405.1
XM_011532881.2 2558 Missense Mutation TCA,TTA S707L XP_011531183.1
XM_011532884.2 2558 Intron XP_011531186.1
XM_011532886.2 2558 Intron XP_011531188.1
XM_011532887.2 2558 Intron XP_011531189.1
XM_011532890.2 2558 Intron XP_011531192.1
XM_011532891.2 2558 Intron XP_011531193.1
XM_017004251.1 2558 Missense Mutation TCA,TTA S707L XP_016859740.1
XM_017004252.1 2558 UTR 3 XP_016859741.1
XM_017004253.1 2558 Intron XP_016859742.1
XM_017004254.1 2558 Intron XP_016859743.1

View Full Product Details