Product Details

SNP ID
rs1064285
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:6102114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTCCCTCCACACCCCTGAACTCA[A/G]ACCCCACGGGCCAGTGGGGACCCTC
Phenotype
MIM: 610771 MIM: 601142
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHD5 PubMed Links

Gene Details

Gene
CHD5
Gene Name
chromodomain helicase DNA binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015557.2 9336 UTR 3 NP_056372.1
Gene
KCNAB2
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199860.1 9336 Intron NP_001186789.1
NM_001199861.1 9336 Intron NP_001186790.1
NM_001199862.1 9336 Intron NP_001186791.1
NM_001199863.1 9336 Intron NP_001186792.1
NM_003636.3 9336 Intron NP_003627.1
NM_172130.2 9336 Intron NP_742128.1
XM_005263514.2 9336 Intron XP_005263571.1
XM_011542321.2 9336 Intron XP_011540623.1
XM_011542322.2 9336 Intron XP_011540624.1
XM_017002618.1 9336 Intron XP_016858107.1
XM_017002619.1 9336 Intron XP_016858108.1
XM_017002620.1 9336 Intron XP_016858109.1
XM_017002621.1 9336 Intron XP_016858110.1

View Full Product Details