Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022037.2 | 4418 | UTR 3 | NP_071320.2 | ||
NM_022173.2 | 4418 | UTR 3 | NP_071505.2 | ||
XM_005264527.1 | 4418 | UTR 3 | XP_005264584.1 | ||
XM_005264528.4 | 4418 | UTR 3 | XP_005264585.1 | ||
XM_005264531.1 | 4418 | UTR 3 | XP_005264588.1 | ||
XM_011533077.1 | 4418 | UTR 3 | XP_011531379.1 | ||
XM_011533078.1 | 4418 | UTR 3 | XP_011531380.1 | ||
XM_011533081.2 | 4418 | UTR 3 | XP_011531383.1 | ||
XM_011533082.2 | 4418 | UTR 3 | XP_011531384.1 | ||
XM_017004789.1 | 4418 | UTR 3 | XP_016860278.1 | ||
XM_017004790.1 | 4418 | UTR 3 | XP_016860279.1 | ||
XM_017004791.1 | 4418 | UTR 3 | XP_016860280.1 | ||
XM_017004792.1 | 4418 | UTR 3 | XP_016860281.1 | ||
XM_017004793.1 | 4418 | UTR 3 | XP_016860282.1 | ||
XM_017004794.1 | 4418 | UTR 3 | XP_016860283.1 | ||
XM_017004795.1 | 4418 | UTR 3 | XP_016860284.1 | ||
XM_017004796.1 | 4418 | UTR 3 | XP_016860285.1 | ||
XM_017004797.1 | 4418 | UTR 3 | XP_016860286.1 | ||
XM_017004798.1 | 4418 | UTR 3 | XP_016860287.1 | ||
XM_017004799.1 | 4418 | UTR 3 | XP_016860288.1 | ||
XM_017004800.1 | 4418 | UTR 3 | XP_016860289.1 |