Product Details

SNP ID
rs1062884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14905631 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACATTTAAGTCCTGGAACTCTT[A/C]TCCAAGATAAATGTGGCTGACTGCT
Phenotype
MIM: 605988 MIM: 606503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCLRE1C PubMed Links

Gene Details

Gene
DCLRE1C
Gene Name
DNA cross-link repair 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033855.2 5409 UTR 3 NP_001029027.1
NM_001033857.2 5409 UTR 3 NP_001029029.1
NM_001033858.2 5409 UTR 3 NP_001029030.1
NM_001289076.1 5409 UTR 3 NP_001276005.1
NM_001289077.1 5409 UTR 3 NP_001276006.1
NM_001289078.1 5409 UTR 3 NP_001276007.1
NM_001289079.1 5409 UTR 3 NP_001276008.1
NM_022487.3 5409 UTR 3 NP_071932.2
XM_006717491.3 5409 Intron XP_006717554.1
XM_011519616.1 5409 Intron XP_011517918.1
XM_011519617.1 5409 Intron XP_011517919.1
XM_011519619.1 5409 Intron XP_011517921.1
XM_011519620.2 5409 Intron XP_011517922.1
XM_011519621.1 5409 Intron XP_011517923.1
XM_017016556.1 5409 Intron XP_016872045.1
XM_017016557.1 5409 Intron XP_016872046.1
XM_017016558.1 5409 Intron XP_016872047.1
XM_017016559.1 5409 Intron XP_016872048.1
XM_017016560.1 5409 Intron XP_016872049.1
Gene
SUV39H2
Gene Name
suppressor of variegation 3-9 homolog 2
There are no transcripts associated with this gene.

View Full Product Details