Product Details
- SNP ID
-
rs911119
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:23632100 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GAGGGGAGGCAGGGGTGGCACCAAC[C/T]TGCTTCCCTGTGGGCTGTGGGCCAT
- Phenotype
-
MIM: 604312
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CST3
PubMed Links
Gene Details
- Gene
- CST3
- Gene Name
- cystatin C
View Full Product Details