Product Details
- SNP ID
-
rs1010933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:76505502 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCACAAGACAGAGATGGACCGCAA[C/T]ATTACGGGCCACGGCTATCCCGACC
- Phenotype
-
MIM: 613141
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DTX2
PubMed Links
Gene Details
- Gene
- DTX2
- Gene Name
- deltex E3 ubiquitin ligase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001102594.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
NP_001096064.1 |
NM_001102595.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
NP_001096065.1 |
NM_001102596.1 |
2326 |
Silent Mutation |
AAC,AAT |
N543N |
NP_001096066.1 |
NM_020892.2 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
NP_065943.2 |
XM_005250131.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_005250188.1 |
XM_005250132.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_005250189.1 |
XM_011515772.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_011514074.1 |
XM_011515773.2 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_011514075.1 |
XM_011515774.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_011514076.1 |
XM_011515775.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_011514077.1 |
XM_017011723.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_016867212.1 |
XM_017011724.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_016867213.1 |
XM_017011725.1 |
2326 |
Silent Mutation |
AAC,AAT |
N590N |
XP_016867214.1 |
XM_017011726.1 |
2326 |
Silent Mutation |
AAC,AAT |
N543N |
XP_016867215.1 |
XM_017011727.1 |
2326 |
Silent Mutation |
AAC,AAT |
N543N |
XP_016867216.1 |
XM_017011728.1 |
2326 |
Silent Mutation |
AAC,AAT |
N536N |
XP_016867217.1 |
XM_017011729.1 |
2326 |
Intron |
|
|
XP_016867218.1 |
XM_017011730.1 |
2326 |
Intron |
|
|
XP_016867219.1 |
- Gene
- LOC105375355
- Gene Name
- uroplakin-3b
There are no transcripts associated with this gene.
View Full Product Details