Product Details

SNP ID
rs1053725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72003166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCAGGAACCAGGGCCTACTCCT[C/T]TTATGGTCCCTTCTAGATCCAGAGG
Phenotype
MIM: 604113 MIM: 164009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL18BP PubMed Links

Gene Details

Gene
IL18BP
Gene Name
interleukin 18 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039659.1 6914 Intron NP_001034748.1
NM_001039660.1 6914 Intron NP_001034749.1
NM_001145055.1 6914 Intron NP_001138527.1
NM_001145057.1 6914 Intron NP_001138529.1
NM_005699.3 6914 Intron NP_005690.2
NM_173042.2 6914 Intron NP_766630.2
NM_173044.2 6914 Intron NP_766632.2
XM_017017059.1 6914 Intron XP_016872548.1
XM_017017060.1 6914 Intron XP_016872549.1
XM_017017061.1 6914 Intron XP_016872550.1
XM_017017062.1 6914 Intron XP_016872551.1
XM_017017063.1 6914 Intron XP_016872552.1
Gene
NUMA1
Gene Name
nuclear mitotic apparatus protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286561.1 6914 UTR 3 NP_001273490.1
NM_006185.3 6914 UTR 3 NP_006176.2
XM_006718564.1 6914 UTR 3 XP_006718627.1
XM_011545054.1 6914 UTR 3 XP_011543356.1
XM_011545055.1 6914 UTR 3 XP_011543357.1
XM_011545056.2 6914 UTR 3 XP_011543358.1
XM_011545057.2 6914 UTR 3 XP_011543359.1
XM_011545058.1 6914 UTR 3 XP_011543360.1
XM_011545059.2 6914 UTR 3 XP_011543361.1
XM_011545060.2 6914 UTR 3 XP_011543362.1
XM_011545061.2 6914 UTR 3 XP_011543363.1
XM_011545062.2 6914 UTR 3 XP_011543364.1
XM_011545063.2 6914 UTR 3 XP_011543365.1
XM_011545064.1 6914 UTR 3 XP_011543366.1
XM_011545065.1 6914 UTR 3 XP_011543367.1
XM_011545066.1 6914 UTR 3 XP_011543368.1
XM_017017830.1 6914 Intron XP_016873319.1
XM_017017831.1 6914 Intron XP_016873320.1
Gene
RNF121
Gene Name
ring finger protein 121
There are no transcripts associated with this gene.

View Full Product Details