Product Details
- SNP ID
-
rs1541304
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:72003032 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTCCCCCAACCCCACTCCTGAGACA[C/T]AGGGCCCATCCGTCCTGGGAAAGAG
- Phenotype
-
MIM: 604113
MIM: 164009
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL18BP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5743679] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL18BP
- Gene Name
- interleukin 18 binding protein
- Gene
- NUMA1
- Gene Name
- nuclear mitotic apparatus protein 1
- Gene
- RNF121
- Gene Name
- ring finger protein 121
There are no transcripts associated with this gene.
View Full Product Details