Product Details

SNP ID
rs1346925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72818006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTGCCAGGCCCTGACCTGAAG[A/C]TGAGCAGGGGTCAGAGTGCCCTGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG16L2 PubMed Links

Gene Details

Gene
ATG16L2
Gene Name
autophagy related 16 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318766.1 Intron NP_001305695.1
NM_033388.1 Intron NP_203746.1

View Full Product Details