Product Details

SNP ID
rs1795615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:64611285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCGTGGGCCTCGCCACCATAGGC[G/T]CTCACTAAATCTCCGTTGCGGGACT
Phenotype
MIM: 607019
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RASSF3 PubMed Links

Gene Details

Gene
RASSF3
Gene Name
Ras association domain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178169.3 Intron NP_835463.1
XM_006719345.3 Intron XP_006719408.1
XM_011538195.2 Intron XP_011536497.1
XM_017019182.1 Intron XP_016874671.1

View Full Product Details