Product Details

SNP ID
rs1209511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:99983800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTTCAAATCTTATTTGAAACAT[C/T]ACTTCCTCAGGGAGGCAGCCTTTGA
Phenotype
MIM: 616912
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVL PubMed Links

Gene Details

Gene
EVL
Gene Name
Enah/Vasp-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016337.2 Intron NP_057421.1
XM_005267749.3 Intron XP_005267806.1
XM_011536828.2 Intron XP_011535130.1
XM_017021363.1 Intron XP_016876852.1
XM_017021364.1 Intron XP_016876853.1

View Full Product Details