Product Details

SNP ID
rs1064761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2833110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAATCCTGCAGAACAAGGGACA[A/G]AAAAGTCAGGCGAGGCAAGCTTGTC
Phenotype
MIM: 130135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB1 PubMed Links

Gene Details

Gene
SERPINB1
Gene Name
serpin family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030666.3 2018 UTR 3 NP_109591.1
XM_011514333.1 2018 UTR 3 XP_011512635.1
XM_011514334.2 2018 UTR 3 XP_011512636.1
XM_011514335.2 2018 UTR 3 XP_011512637.1

View Full Product Details