Product Details

SNP ID
rs1152207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108048961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAGGTAAAACCAAAATAAATGC[G/T]TGTAGCACTCAATATCTCAAACCTA
Phenotype
MIM: 601028
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD47 PubMed Links

Gene Details

Gene
CD47
Gene Name
CD47 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001777.3 Intron NP_001768.1
NM_198793.2 Intron NP_942088.1
XM_005247908.1 Intron XP_005247965.1
XM_005247909.1 Intron XP_005247966.1
XM_017007536.1 Intron XP_016863025.1

View Full Product Details