Product Details

SNP ID
rs1150932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27447896 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGGACTTACGCGTCTGAGCCCCA[C/G]AAACTCAGCCGAGTGCCTCTCCTAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARNTL2-AS1 PubMed Links

Gene Details

Gene
ARNTL2-AS1
Gene Name
ARNTL2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SMCO2
Gene Name
single-pass membrane protein with coiled-coil domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145010.1 Intron NP_001138482.1
XM_011520634.2 Intron XP_011518936.1
XM_011520635.2 Intron XP_011518937.1
XM_011520636.2 Intron XP_011518938.1
XM_017019254.1 Intron XP_016874743.1
XM_017019255.1 Intron XP_016874744.1

View Full Product Details