Product Details
- SNP ID
-
rs1552463
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:7241712 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATAGTCTGTAGTAAATCACCTGGG[C/T]TTTAATCAATTCTTTAGTAAGCTGT
- Phenotype
-
MIM: 613528
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR302E
PubMed Links
Gene Details
- Gene
- MIR302E
- Gene Name
- microRNA 302e
There are no transcripts associated with this gene.
- Gene
- SYT9
- Gene Name
- synaptotagmin 9
View Full Product Details