Product Details

SNP ID
rs1760943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20458540 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCCACCAGGGAGGGGCCAGGCAC[A/G]GTCTGTGGGTCATCAGGCTCAGGAG
Phenotype
MIM: 107748 MIM: 610107 MIM: 609865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244249.1 1161 Intron NP_001231178.1
NM_001641.3 1161 Intron NP_001632.2
NM_080648.2 1161 Intron NP_542379.1
NM_080649.2 1161 Intron NP_542380.1
Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
There are no transcripts associated with this gene.

Gene
TMEM55B
Gene Name
transmembrane protein 55B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100814.2 1161 UTR 3 NP_001094284.1
NM_144568.3 1161 UTR 3 NP_653169.2

View Full Product Details