Product Details

SNP ID
rs859603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:129781102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCAAGTATATCCTACACATCCA[A/G]TGGGATGCTAGGACAGTCCCAAAGC
Phenotype
MIM: 300441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SASH3 PubMed Links

Gene Details

Gene
SASH3
Gene Name
SAM and SH3 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018990.3 Intron NP_061863.1
XM_006724763.1 Intron XP_006724826.1

View Full Product Details