Product Details

SNP ID
rs854189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45391062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGTAGCTAATTGCTTGCTTTCC[A/C]ATTCTGTTCTTAATGGTTTTGATAT
Phenotype
MIM: 604544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LARS2 PubMed Links

Gene Details

Gene
LARS2
Gene Name
leucyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015340.3 Intron NP_056155.1
XM_005265006.1 Intron XP_005265063.1
XM_011533554.2 Intron XP_011531856.1
XM_017006042.1 Intron XP_016861531.1

View Full Product Details