Product Details

SNP ID
rs1264306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30914426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTTGGGATGGGCGGGAAGGC[C/T]TGGCCGCGCCGGGCTGTGGGCACTG
Phenotype
MIM: 601760 MIM: 612802
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTF2H4 PubMed Links

Gene Details

Gene
GTF2H4
Gene Name
general transcription factor IIH subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001517.4 96 Intron NP_001508.1
Gene
VARS2
Gene Name
valyl-tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167733.2 96 Intron NP_001161205.1
NM_001167734.1 96 Silent Mutation GCC,GCT A5A NP_001161206.1
NM_020442.5 96 Intron NP_065175.4

View Full Product Details