Product Details

SNP ID
rs13305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128258722 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGCTACAGTGGTTGTGCTTCA[A/T]CTTCTGTTGCCTGGTGTGTCTGGTG
Phenotype
MIM: 612570
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FBN2 PubMed Links

Gene Details

Gene
FBN2
Gene Name
fibrillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001999.3 10074 UTR 3 NP_001990.2
XM_017009228.1 10074 UTR 3 XP_016864717.1

View Full Product Details