Product Details
- SNP ID
-
rs1801171
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:128259452 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAGCCTGGGCCCAAGTCTGTGAA[C/G]GGTTAATAGAGCTGAATCTGCAGCC
- Phenotype
-
MIM: 612570
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBN2
PubMed Links
Gene Details
- Gene
- FBN2
- Gene Name
- fibrillin 2
View Full Product Details