Product Details

SNP ID
rs1560680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149973554 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTCTCCCCTCATCTCTCTCTCCC[A/G]TTCCCCCCACATATATATGGTGCTG
Phenotype
MIM: 606718
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC26A2 PubMed Links

Gene Details

Gene
SLC26A2
Gene Name
solute carrier family 26 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000112.3 Intron NP_000103.2
XM_017009191.1 Intron XP_016864680.1

View Full Product Details