Product Details

SNP ID
rs1422159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170785387 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGATTTGACTCTAAAAATTAACA[C/G]CTTCTGGGCCAGCAGCCATGAGAAT
Phenotype
MIM: 602729
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GABRP PubMed Links

Gene Details

Gene
GABRP
Gene Name
gamma-aminobutyric acid type A receptor pi subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291985.1 Intron NP_001278914.1
NM_014211.2 Intron NP_055026.1
XM_005265872.2 Intron XP_005265929.1

View Full Product Details