Product Details

SNP ID
rs8764
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:7981425 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAACAACTATATTTTGGACAAAAC[A/C]ATTTTTTTTTAATCTGTCTTGTAAA
Phenotype
MIM: 616766
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
TMX4 PubMed Links
Additional Information
For this assay, SNP(s) [rs200269615] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TMX4
Gene Name
thioredoxin related transmembrane protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021156.3 2092 UTR 3 NP_066979.2

View Full Product Details