Product Details
- SNP ID
-
rs3815844
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:231062855 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGATCTGCCCTTCAACTGAGTGCT[G/T]GTTCTCAGTGGTTATGATGATCACC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PSMD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74757457] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PSMD1
- Gene Name
- proteasome 26S subunit, non-ATPase 1
View Full Product Details