Product Details
- SNP ID
-
rs62527225
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:85251634 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAACCTATAATGCCATTTAAGTAC[A/C]TGCTTTAATAGAAAAAGTTTAGTTT
- Phenotype
-
MIM: 611436
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CA13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75647642] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CA13
- Gene Name
- carbonic anhydrase 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198584.2 |
|
Intron |
|
|
NP_940986.1 |
View Full Product Details