Product Details

SNP ID
rs4874105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143375158 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGGGGCAAGGGGGGGATCCAGA[A/G]TGAAATCCAGAAGCGCAGAAGGAAG
Phenotype
MIM: 601031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RHPN1 PubMed Links

Gene Details

Gene
RHPN1
Gene Name
rhophilin Rho GTPase binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052924.2 Intron NP_443156.2
XM_005250772.3 Intron XP_005250829.1
XM_005250773.3 Intron XP_005250830.1
XM_006716502.3 Intron XP_006716565.1
XM_011516817.2 Intron XP_011515119.1
XM_011516818.1 Intron XP_011515120.1
XM_011516819.2 Intron XP_011515121.1
XM_011516820.2 Intron XP_011515122.1
XM_011516821.2 Intron XP_011515123.1
XM_011516822.2 Intron XP_011515124.1
XM_011516823.2 Intron XP_011515125.1
XM_017013013.1 Intron XP_016868502.1
XM_017013014.1 Intron XP_016868503.1
XM_017013015.1 Intron XP_016868504.1
XM_017013016.1 Intron XP_016868505.1
Gene
RHPN1-AS1
Gene Name
RHPN1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details