Product Details

SNP ID
rs10086425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:51822774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCGTGACATGCAAAGACCAAGA[A/G]TAGAAGAAAGCACCCAGAACTAAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCMTD1 PubMed Links

Gene Details

Gene
PCMTD1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286782.1 Intron NP_001273711.1
NM_001286783.1 Intron NP_001273712.1
NM_052937.3 Intron NP_443169.2
XM_005251146.2 Intron XP_005251203.1
XM_011517444.2 Intron XP_011515746.1
XM_017013017.1 Intron XP_016868506.1

View Full Product Details