Product Details

SNP ID
rs77680937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99717215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTATTAGCAGGTTTTGAAAGCT[A/G]GTTGAGTCCTGTGGCTAAAAATACT
Phenotype
MIM: 600660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYSMD4 PubMed Links

Gene Details

Gene
LYSMD4
Gene Name
LysM domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284417.1 1125 Intron NP_001271346.1
NM_001284418.1 1125 Intron NP_001271347.1
NM_001284419.1 1125 Intron NP_001271348.1
NM_001284420.1 1125 Intron NP_001271349.1
NM_001284421.1 1125 Intron NP_001271350.1
NM_001284422.1 1125 Intron NP_001271351.1
NM_152449.3 1125 Intron NP_689662.2
XM_006720397.3 1125 Intron XP_006720460.2
XM_011521241.1 1125 Intron XP_011519543.1
XM_011521242.2 1125 UTR 3 XP_011519544.1
XM_011521243.2 1125 Intron XP_011519545.1
XM_011521244.1 1125 Intron XP_011519546.1
XM_011521245.2 1125 UTR 3 XP_011519547.1
XM_017021926.1 1125 Intron XP_016877415.1
XM_017021927.1 1125 Intron XP_016877416.1
XM_017021928.1 1125 Intron XP_016877417.1
XM_017021929.1 1125 Intron XP_016877418.1
XM_017021930.1 1125 Intron XP_016877419.1
Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 1125 Intron NP_001124398.1
NM_001130927.2 1125 Intron NP_001124399.1
NM_001130928.2 1125 Intron NP_001124400.1
NM_001171894.2 1125 Intron NP_001165365.1
NM_001319206.1 1125 Intron NP_001306135.1
NM_005587.3 1125 Intron NP_005578.2
XM_005254914.2 1125 Intron XP_005254971.1
XM_005254915.2 1125 Intron XP_005254972.1
XM_005254916.3 1125 Intron XP_005254973.1
XM_011521571.2 1125 Intron XP_011519873.1
XM_011521572.2 1125 Intron XP_011519874.1
XM_011521573.2 1125 Intron XP_011519875.1
XM_011521576.2 1125 Intron XP_011519878.1
XM_011521577.2 1125 Intron XP_011519879.1
XM_011521578.2 1125 Intron XP_011519880.1
XM_011521579.2 1125 Intron XP_011519881.1
XM_011521581.2 1125 Intron XP_011519883.1
XM_011521582.2 1125 Intron XP_011519884.1
XM_011521583.2 1125 Intron XP_011519885.1
XM_011521585.2 1125 Intron XP_011519887.1
XM_011521586.2 1125 Intron XP_011519888.1
XM_011521587.2 1125 Intron XP_011519889.1
XM_011521590.2 1125 Intron XP_011519892.1
XM_017022190.1 1125 Intron XP_016877679.1
XM_017022191.1 1125 Intron XP_016877680.1
XM_017022192.1 1125 Intron XP_016877681.1
XM_017022193.1 1125 Intron XP_016877682.1
XM_017022194.1 1125 Intron XP_016877683.1
XM_017022195.1 1125 Intron XP_016877684.1
XM_017022196.1 1125 Intron XP_016877685.1
XM_017022197.1 1125 Intron XP_016877686.1
XM_017022198.1 1125 Intron XP_016877687.1
XM_017022199.1 1125 Intron XP_016877688.1
XM_017022200.1 1125 Intron XP_016877689.1
XM_017022201.1 1125 Intron XP_016877690.1
XM_017022202.1 1125 Intron XP_016877691.1
XM_017022203.1 1125 Intron XP_016877692.1
XM_017022204.1 1125 Intron XP_016877693.1

View Full Product Details