Product Details

SNP ID
rs28641162
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:50713822 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTCAATGAGTCGAGAAATAACCAAA[C/T]CAACAAACAACAAAACAAAAAATCC
Phenotype
MIM: 608238
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPPL2A PubMed Links

Gene Details

Gene
SPPL2A
Gene Name
signal peptide peptidase like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032802.3 Intron NP_116191.2
XM_005254722.3 Intron XP_005254779.1
XM_017022680.1 Intron XP_016878169.1
XM_017022681.1 Intron XP_016878170.1

View Full Product Details