Product Details

SNP ID
rs770911
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:82364648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCCTCAGAGGATCCAGCGAGGGG[C/T]GCCAACAAGAGGCGAAGAGGTGGCA
Phenotype
MIM: 190920
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TPBG PubMed Links

Gene Details

Gene
TPBG
Gene Name
trophoblast glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166392.1 112 UTR 5 NP_001159864.1
NM_006670.4 112 UTR 5 NP_006661.1
XM_011536097.2 112 UTR 5 XP_011534399.1

View Full Product Details