Product Details

SNP ID
rs1013559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34674780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGTTCTTTTTCTTTCCTCATAA[A/T]AACAAAGGGTAACAACATAATCACA
Phenotype
MIM: 615321
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLIC6 PubMed Links

Gene Details

Gene
CLIC6
Gene Name
chloride intracellular channel 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317009.1 Intron NP_001303938.1
NM_053277.2 Intron NP_444507.1
XM_017028406.1 Intron XP_016883895.1

View Full Product Details