Product Details

SNP ID
rs875224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:36703013 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGATTAACCCTTCATACCTGGG[A/G]AAATTAAGGCCAGACATGACACAGG
Phenotype
MIM: 600892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SIM2 PubMed Links

Gene Details

Gene
SIM2
Gene Name
single-minded family bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005069.4 Intron NP_005060.1
NM_009586.3 Intron NP_033664.2
XM_011529694.1 Intron XP_011527996.1
XM_017028442.1 Intron XP_016883931.1

View Full Product Details