Product Details
- SNP ID
-
rs1143641
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112831089 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACTATGGTAGGACATAATAACATC[A/G]GCAAAATGATTTAATTTTCTGCAGC
- Phenotype
-
MIM: 147720
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL1B
PubMed Links
Gene Details
- Gene
- IL1B
- Gene Name
- interleukin 1 beta
View Full Product Details