Product Details

SNP ID
rs1743180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91581754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTGAGATGTGAGGCAAGGGTG[A/G]GGTCATTGTATGTTAAATATAGAAG
Phenotype
MIM: 611169
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPERB PubMed Links

Gene Details

Gene
CATSPERB
Gene Name
cation channel sperm associated auxiliary subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024764.3 Intron NP_079040.2

View Full Product Details