Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000295.4 | 2644 | UTR 3 | NP_000286.3 | ||
NM_001002235.2 | 2644 | UTR 3 | NP_001002235.1 | ||
NM_001002236.2 | 2644 | UTR 3 | NP_001002236.1 | ||
NM_001127700.1 | 2644 | UTR 3 | NP_001121172.1 | ||
NM_001127701.1 | 2644 | UTR 3 | NP_001121173.1 | ||
NM_001127702.1 | 2644 | UTR 3 | NP_001121174.1 | ||
NM_001127703.1 | 2644 | UTR 3 | NP_001121175.1 | ||
NM_001127704.1 | 2644 | UTR 3 | NP_001121176.1 | ||
NM_001127705.1 | 2644 | UTR 3 | NP_001121177.1 | ||
NM_001127706.1 | 2644 | UTR 3 | NP_001121178.1 | ||
NM_001127707.1 | 2644 | UTR 3 | NP_001121179.1 | ||
XM_017021370.1 | 2644 | Intron | XP_016876859.1 |