Product Details

SNP ID
rs12836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46956869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAAAGCTTTGTGGTTAGTTTAAA[A/T]TATACACTCTGTAGATACTATACCA
Phenotype
MIM: 611322
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA2 PubMed Links

Gene Details

Gene
DNAJA2
Gene Name
DnaJ heat shock protein family (Hsp40) member A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005880.3 1541 UTR 3 NP_005871.1

View Full Product Details