Product Details

SNP ID
rs711752
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56962299 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCCTTCAAGGTCAAGTTCTTTGGT[A/G]AGAAGGTCCTAGCTGCATTGCAAAC
Phenotype
MIM: 118470
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000078.2 Intron NP_000069.2
NM_001286085.1 Intron NP_001273014.1
XM_006721124.3 Intron XP_006721187.1

View Full Product Details