Product Details

SNP ID
rs794455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7895031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGTCCCTGCCCCTGGGGCCCT[A/G]GTAACCAGAGCCAGGCTCCAGCCGG
Phenotype
MIM: 612891 MIM: 603014
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC8E PubMed Links

Gene Details

Gene
LRRC8E
Gene Name
leucine rich repeat containing 8 family member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001268284.2 516 Intron NP_001255213.1
NM_001268285.2 516 Intron NP_001255214.1
NM_025061.5 516 Intron NP_079337.2
XM_011528319.2 516 UTR 5 XP_011526621.1
Gene
MAP2K7
Gene Name
mitogen-activated protein kinase kinase 7
There are no transcripts associated with this gene.

View Full Product Details