Product Details

SNP ID
rs1467512
Assay Type
Functionally tested
NCBI dbSNP Submissions
39
Location
Chr.1:229626529 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
AGCAGACTCTGGGCTGGAGCGGCTT[C/G]GGCACGCTCCATTCGCGAAGGCGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TAF5L PubMed Links

Gene Details

Gene
TAF5L
Gene Name
TATA-box binding protein associated factor 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025247.1 Intron NP_001020418.1
NM_014409.3 Intron NP_055224.1
XM_005273099.4 Intron XP_005273156.1
XM_005273100.2 Intron XP_005273157.1
XM_011544162.1 Intron XP_011542464.1
XM_011544163.1 Intron XP_011542465.1
XM_011544164.2 Intron XP_011542466.1
XM_017001045.1 Intron XP_016856534.1
Gene
URB2
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314021.1 Intron NP_001300950.1
NM_014777.3 Intron NP_055592.2
XM_005273360.3 Intron XP_005273417.1

View Full Product Details