Product Details

SNP ID
rs1059648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:57600800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTTAATAAGGTAGCACAGATTA[A/G]ATCTAAGGACATGGTGGGGGCAACT
Phenotype
MIM: 108410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NARS PubMed Links

Gene Details

Gene
NARS
Gene Name
asparaginyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004539.3 2727 UTR 3 NP_004530.1
XM_005266700.1 2727 UTR 3 XP_005266757.1

View Full Product Details