Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001184866.1 | 143 | UTR 5 | NP_001171795.1 | ||
NM_001184867.1 | 143 | UTR 5 | NP_001171796.1 | ||
NM_001184870.1 | 143 | UTR 5 | NP_001171799.1 | ||
NM_001184871.1 | 143 | UTR 5 | NP_001171800.1 | ||
NM_001184872.1 | 143 | UTR 5 | NP_001171801.1 | ||
NM_001184873.1 | 143 | UTR 5 | NP_001171802.1 | ||
NM_032738.3 | 143 | UTR 5 | NP_116127.3 | ||
XM_006711581.3 | 143 | UTR 5 | XP_006711644.1 | ||
XM_011510064.1 | 143 | Intron | XP_011508366.1 | ||
XM_011510065.1 | 143 | Intron | XP_011508367.1 | ||
XM_011510066.1 | 143 | UTR 5 | XP_011508368.1 |