Product Details

SNP ID
rs1334849
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:153032176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGAGCAAGAGCAAATCATCTTTA[A/G]GTTCCTCTTCTTCAAAGGCTCAGAA
Phenotype
MIM: 182266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR1B PubMed Links

Gene Details

Gene
SPRR1B
Gene Name
small proline rich protein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003125.2 Intron NP_003116.2
Gene
SPRR2D
Gene Name
small proline rich protein 2D
There are no transcripts associated with this gene.

View Full Product Details