Product Details

SNP ID
rs9826
Assay Type
Functionally tested
NCBI dbSNP Submissions
68
Location
Chr.1:151806423 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCCTCCAGGATCTGATCTTGCC[C/T]TCCGACTGACCAGTGCGAGCGACTT
Phenotype
MIM: 609794 MIM: 602943
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINGO4 PubMed Links

Gene Details

Gene
LINGO4
Gene Name
leucine rich repeat and Ig domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004432.3 3497 Intron NP_001004432.1
Gene
RORC
Gene Name
RAR related orphan receptor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001523.1 3497 UTR 3 NP_001001523.1
NM_005060.3 3497 UTR 3 NP_005051.2
XM_006711484.3 3497 UTR 3 XP_006711547.2

View Full Product Details