Product Details

SNP ID
rs1131473
Assay Type
Validated
NCBI dbSNP Submissions
56
Location
Chr.1:152107347 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCAGTGTTTCTCATTTTCCCGTGCT[C/G]GAAGCTTTGGCAGGTGTCAAGATAT
Phenotype
MIM: 190370
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCHH PubMed Links

Gene Details

Gene
TCHH
Gene Name
trichohyalin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007113.3 5965 UTR 3 NP_009044.2

View Full Product Details