Product Details
- SNP ID
-
rs872180
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:152776852 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCAAACTCTGTCCTGGAAGATTC[C/T]TCCGACCTAGAATCCAGAAATCTGC
- Phenotype
-
MIM: 612607
MIM: 612608
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LCE1E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115176407] are located under a probe and SNP(s) [rs80342527] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LCE1E
- Gene Name
- late cornified envelope 1E
There are no transcripts associated with this gene.
- Gene
- LCE1F
- Gene Name
- late cornified envelope 1F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178354.2 |
481 |
UTR 3 |
|
|
NP_848131.1 |
View Full Product Details