Product Details

SNP ID
rs4772
Assay Type
Validated
NCBI dbSNP Submissions
59
Location
Chr.1:153373787 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGTGAGGGAAAGAAAAGACCCTC[A/G]TTGAGGACATTGCTGGGTAAAAAGC
Phenotype
MIM: 603112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A12 PubMed Links

Gene Details

Gene
S100A12
Gene Name
S100 calcium binding protein A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005621.1 387 UTR 3 NP_005612.1

View Full Product Details