Product Details

SNP ID
rs822522
Assay Type
Functionally tested
NCBI dbSNP Submissions
78
Location
Chr.1:155868074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACCAGCAGGCAGAGAAGAAGCA[C/G]AAGAACCCACCATACAAGTTTATTC
Phenotype
MIM: 610393 MIM: 608741
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GON4L PubMed Links

Gene Details

Gene
GON4L
Gene Name
gon-4 like
There are no transcripts associated with this gene.

Gene
SYT11
Gene Name
synaptotagmin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152280.4 452 Missense Mutation CAC,CAG H48Q NP_689493.3
XM_005245014.2 452 Missense Mutation CAC,CAG H48Q XP_005245071.1
XM_017000759.1 452 Missense Mutation CAC,CAG H48Q XP_016856248.1

View Full Product Details